missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lectin HPA From Helix pomatia (edible snail), Alexa Fluor™ 488 Conjugate
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Lectin Helix pomatia agglutinin (HPA) selectively binds to α-N-acetylgalactosamine residues and type A erythrocytes.
Cell Analysis, Cellular Imaging, Flow Cytometry, Flow Cytometry Antibodies & Secondary Detection, Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Quantity | 1 mg |
| Source | Edible snail |
| Conjugate | Alexa Fluor 488 |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Label Type | Alexa Fluor Dyes |
| Shipping Condition | Room Temperature |
| Form | Lyophilized |
| Protein Subtype | Lectins |
| Product Line | Alexa Fluor |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction