missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lectin GS-II From Griffonia simplicifolia, Alexa Fluor™ 647 Conjugate

Product Code. 11514976
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11514976

Brand: Invitrogen™ L32451

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Isolated from the seeds of Griffonia simplicifolia, a tropical African legume

Lectin GS-II is a 113,000-dalton compoased of identical subunits. Each subunit contains a single binding site specific for terminal non-reducing a- or Β-linked N-acetyl-D-glucoasemine. Unlike many other lectins, GS-II does not react with any specific blood group. The far red-fluorescent lectin GS-II Alexa Fluor 647 conjugate can be used in fluorescence microscopy or flow cytometry.

Cell Analysis, Cell Tracing & Tracking, Cellular Imaging, Flow Cytometry, Flow Cytometry Antibodies & Secondary Detection, General Cell Tracing, Immunofluorescence (IF), Immunofluorescence Staining & Detection, In Vivo Imaging, Microbial Tracking

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 500 μg
Source Africa Shrub legume
Conjugate Alexa Fluor 647
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label Type Alexa Fluor Dyes
Shipping Condition Room Temperature
Form Lyophilized
Protein Subtype Lectins
Product Line Alexa Fluor

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.