missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ImaGene Red™ C12RG lacZ Gene Expression Kit

Product Code. 10709774
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10709774 1 kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10709774 Supplier Invitrogen™ Supplier No. I2906
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Substrates cleaved within cell for fluorescent product

The ImaGene Red™ lacZ Gene Expression Kit contains a fluorescein-based galactosidase substrate that has been covalently modified to include a 12-carbon lipophilic moiety, C12RG. Once inside the cell, the substrates are cleaved by β-galactosidase, producing a fluorescent product that is well retained by the cells, probably by incorporation of the lipophilic tail within the cellular membrane. In addition to the C12RG, the kits also include the broad-spectrum β-galactosidase inhibitor, PETG, and chloroquine diphosphate for inhibiting acidic hydrolysis of the substrate.

Beta-Galactosidase Assays, Cell Analysis, Cellular Imaging, Enzyme and Protein Activity Assays, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Proteins, Expression, Isolation and Analysis, Reporter Assays

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Product Type β-Galactosidase Reporter Gene Assay System
Detection Method Fluorescence
Assay β-Gal (lacZ) Assay
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Shipping Condition Room Temperature
Substrate C12RG
Substrate Type Beta-Gal Substrate
Substrate Properties Chemical Substrate
Target Enzyme Beta-Galactosidase
Format Kit
Product Line DetectaGene
Quantity 1 kit
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.