missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FocalCheck™ Microspheres, 6 μm, fluorescent dark-red ring stain/green throughout

Product Code. 10575393
missing translation for 'orderingAttributeHoverText'
Quantity:
0.5 mL
missing translation for 'unitSize'
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10575393

missing translation for 'mfr': Invitrogen™ F14807

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Fluorescent green/orange/dark-red ring stains

The sharp ring stains exhibited by the 6.0μm FocalCheck fluorescent green/orange/dark-red ring stained microspheres produce a striking visual representation of instrument misalignment or other aberrations making them ideal as reference standards for confocal laser-scanning microscopy. Correct image registration is indicated when the multiple ring images are perfectly coincident in all dimensions.

Cell Analysis, Cell Cycle, Cell Viability, Proliferation and Function, Cellular Imaging, Immunofluorescence (IF)

TRUSTED_SUSTAINABILITY

Specifications

Calibration Type Confocal Microscope Calibration
Content And Storage Store in refrigerator 2°C to 8°C and protect from light.
Color Dark Red, Green
Product Type Microsphere
Format Suspension Beads
Product Line FocalCheck
Quantity 0.5 mL
Shipping Condition Room Temperature
Diameter (Metric) 6 μm

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.