missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FocalCheck™ Fluorescence Microscope Test Slide #3, 5 colors, high & low intensities

Product Code. 10063572
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10063572 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10063572 Supplier Invitrogen™ Supplier No. F36914

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Five colors, high and low intensities for confocal and fluorescence microscope calibration

The FocalCheck fluorescence microscope test slide #3 is specifically designed for calibrating fluorescence microscope systems and evaluating system and filter performance. This slide is useful for basic evaluation of filter performance and as a general practice slide for fluorescence microscopy and digital imaging.

Cell Analysis, Cell Cycle, Cell Viability, Proliferation and Function, Cellular Imaging, Immunofluorescence (IF)

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Calibration Type Confocal Microscope Calibration, Fluorescence Microscope Calibration
Content And Storage Store at room temperature.
Color Assorted Colors
Product Type Fluorescence Microscope Test Slide
Format Suspension Beads
Product Line FocalCheck
Quantity 1 Ea.
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.