missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FluoReporter™ Blue Fluorometric dsDNA Quantitation Kit, 200-2,000 assays

Product Code. 10256762
Click to view available options
Quantity:
1 kit
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10256762

Brand: Invitrogen™ F2962

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Provide rotocols and reagents for analyzing cellular NA with blue-fluorescent Hoechst 33258 nucleic acid stain

  • Quantitation of cellular DNA is rapid, and all manipulations can be carried out in microplate wells
  • Cells are lysed by freezing them in distilled water, diluted dye solution is then added to lysed cells, and fluorescence is measured

Biochemical Toxicology Assays, Cell Analysis, Cell Counting and Viability, Cell Counting, Viability, and Cryopreservation, Cell Culture, Cell Proliferation, Cell Viability, Proliferation and Function, Cellular Imaging, Cellular Toxicology Assays, Drug Discovery and Development, High-Content Screening (HCS), Target-Based ADME/Tox Assays

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2–8°C) and protect from light.
Detection Method Fluorescence
Format 96-well Plate
Label or Dye Other Labels or Dyes
For Use With (Equipment) Microplate Reader
Product Line FluoReporter
Product Type dsDNA Quantitation Kit
Quantity 1 kit
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.