missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FilmTracer™ FM™ 1-43 Green Biofilm Cell Stain

Product Code. 10054054
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10054054 1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10054054 Supplier Invitrogen™ Supplier No. F10317

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Lipophilic styryl compounds suitable for wide variety of studies involving plasma membrane and vesiculation

The water-soluble FM dyes are virtually nonfluorescent in aqueous media and are believed to insert into the surface membrane where they become intensely fluorescent. FM 1-43 stain has been used successfully to stain the cell bodies specifically in a complex biofilm mileu, including Pseudomonas aeruginosa, Escherichia coli, Staphylococcus sp., Acidothiobacillus caldus, and Vibrio cholerae.

  • Flow Cytometer Laser Lines: 488
  • Platform: Confocal Microscope, Flow Cytometer, Fluorescence Microscope, Fluorescent Imager, Microplate Reader
  • Label or Dye: FM 1-43/lipid

Analysis of Bacteria, Bacterial Staining & Detection, Bacterial Tracing/Tracking/Morphology, Cell Analysis, Microbiological Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Green
Content And Storage Store at room temperature; Dessicate; Protect from light
Excitation Wavelength Range 472⁄580
Dye Type FM 1-43⁄lipid
For Use With (Equipment) Confocal Microscope, Fluorescence Microscope, Flow Cytometer, Microplate Reader, Fluorescent Imager
Quantity 1 mg
Detection Method Fluorescence
Emission Visible
Form Solid
Product Line FilmTracer, Molecular Probes
Shipping Condition Room Temperature
Label Type Other Labels or Dyes
Product Type Dye
SubCellular Localization Cell Membranes, Lipids
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.