missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fibrinogen From Human Plasma, Oregon Green™ 488 Conjugate

Product Code. 11524836
Click to view available options
:
5mg
This item is not returnable. View return policy

Product Code. 11524836

Brand: Invitrogen™ F7496

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For investigating platelet activation and subsequent fibrinogen binding

Molecular Probes fibrinogen conjugates are prepared by attaching fluorescent dye molecules to purified human fibrinogen (approximately 15 dye molecules for each fibrinogen molecule), purifying the conjugate to remove unreacted dye, and then lyophilizing for storage. Fluorescently labeled fibrinogen has proven to be a valuable tool for investigating platelet activation and subsequent fibrinogen binding. For instance, fluorescein-labeled fibrinogen has been used to detect fibrinogen bound to activated platelets by flow cytometry.

  • Label (Ex/Em): Oregon Green™ 488 (approx. 496/524)
  • Spectrally similar to fluorescein, but the signal is less pH-dependent and more photostable
  • Lyophilized product can be dissolved in buffer (e.g., sodium bicarbonate, pH 8.3) for use
  • Fluorescence is typically detected using fluorescence microscopy or flow cytometry

Cell Adhesion, Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Endocytosis, Exocytosis and Phagocytosis, Immunofluorescence (IF), Immunofluorescence Staining and Detection

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Conjugate Oregon Green 488
Species Human
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Label Type Classic Dyes
Shipping Condition Room Temperature
Protein Subtype Fibrinogen
Product Line Oregon Green

For Research Use Only. Not for human or animal therapeutic or diagnostic use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.