missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ethidium Homodimer-1 (EthD-1)

Product Code. 10184382
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10184382 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10184382 Supplier Invitrogen™ Supplier No. E1169

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

The cell-impermeant viability indicator ethdium homodimer-1 (EthD-1) is a high-affinity nucleic acid stain that is weakly fluorescent until bound to DNA.

The cell-impermeant viability indicator ethdium homodimer-1 (EthD-1) is a high-affinity nucleic acid stain that is weakly fluorescent until bound to DNA and emits red fluorescence (Excitation/emission maxima ∼528/617 nm).

We recommend using high-quality anhydrous DMSO for a stock concentration of 1 to 2 mM in DMSO/H2O 1:4 (v/v).

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Orange
Content And Storage Store in freezer (-5°C to -30°C) and protect from light.
Excitation Wavelength Range 528 nm
Dye Type Cell-Impermeant
For Use With (Application) Cell staining assays
For Use With (Equipment) Fluorescence Microscope
Quantity 1 mg
Detection Method Fluorescence
Description Ethidium Homodimer-1 (EthD-1)
Emission 617 nm
Shipping Condition Room Temperature
Label Type Fluorescent Dye
Product Type Viability Indicator
SubCellular Localization Nucleic Acids, Nucleus
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethidium Homodimer-1 (EthD-1)
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Substances or mixtures which\, in contact with water\, emits flammable gases Category 3
Hazard Statement
  • H261-In contact with water releases flammable gases.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P223-Do not allow contact with water.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 5,5-[Ethylenebis(iminotrimethylene)]bis[3,8-diamino-6-phenylphenanthridinium] dichloride dihydrochlo

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.