Learn More
Invitrogen™ EnzChek™ Protease Assay Kit, red fluorescence, 100-1000 assays
Description
The EnzChek Protease Assay Kit, red fluorescence, is a fast, simple, and direct fluorescence-based assay for detecting metallo-, serine, acid, and sulfhydryl proteases. The accompanying increase in fluorescence, which can be measured with a fluorescence microplate reader, is proportional to protease activity. This EnzChek Protease Assay Kit contains a casein derivative that has been extensively labeled with the red-fluorescent BODIPY TR-X dye, which results in a quenching of the fluorescent dye. Protease-catalyzed hydrolysis releases the highly-fluorescent BODIPY TR-X dye-labeled peptides, allowing for quantitative detection of protease activity in solution. The red-fluorescent BODIPY TR-X dye has excitation and emission spectra similar to those of the Texas Red fluorophore.
- Generalized mechanism allows for measurement of variety of proteases
- Can be used to continuously measure protease kinetics
- Excitation/emission similar to Texas Red™ dye (589/617nm)
Enzyme and Protein Activity Assays, Protease/Peptidase Activity, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
| Detection Method | Fluorescence Intensity |
| For Use With (Application) | Protease Assay |
| For Use With (Equipment) | Fluorescence Microplate Reader |
| Product Type | EnzChek Assay Kit |
| Product Line | EnzChek |
| Quantity | 1 Kit |
| Shipping Condition | Room Temperature |
| Substrate Properties | Protein-Based Substrate |
| Substrate Type | Protease Substrate |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.