missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EnzChek™ Myeloperoxidase (MPO) Activity Assay Kit

Product Code. 10194482
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10194482 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10194482 Supplier Invitrogen™ Supplier No. E33856

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

For measuring MPO activities and high-throughput screening for MPO-specific inhibitors

Myeloperoxidase (MPO) is unique peroxidase that, in addition to its peroxidation activity, also catalyzes conversion of hydrogen peroxide (H2O2) and chloride (Cl-) to hypochlorous acid (HOCl). The EnzChek™ Myeloperoxidase (MPO) Activity Assay Kit provides assays for determination of both chlorination and peroxidation activities of MPO in solution and in cell lysates.

  • For detection of chlorination, kit includes nonfluorescent 3'-(p-aminophenyl) fluorescein (APF), which is selectively cleaved by hypochlorite (-OCl) to yield fluorescein Peroxidation is detected using nonfluorescent AmplexUltraRed reagent (A36006), which is oxidized by H2O2-generated redox intermediates MPO-I and MPO-II to form fluorescent product
  • EnzChek Myeloperoxidase Activity Assay Kit can be used to continuously detect these activities at room temperature over broad dynamic range (1.5 to 200ng/mL)
  • Kit combines speed (30 minutes), sensitivity and mix-and-read convenience

Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Enzyme and Protein Activity Assays, High-Content Screening (HCS), Nitro-Oxidative Stress, Oxidase and Peroxidase Activity, Proteins, Expression, Isolation and Analysis, Reactive Oxygen Species

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator 2°C to 8°C and protect from light.
Detection Method Fluorescence Intensity
For Use With (Application) Myeloperoxidase (MPO) Activity Assay
Product Type EnzChek Assay Kit
Product Line EnzChek
Quantity 1 Kit
Shipping Condition Wet Ice
Substrate Properties Chemical Substrate
Target Enzyme Myeloperoxidase

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.