missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EnzChek™ Ultra Amylase Assay Kit

Product Code. 10093552
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10093552 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10093552 Supplier Invitrogen™ Supplier No. E33651

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Measures amylase activity or screens amylase inhibitors in a high-throughput format

The EnzChek Ultra Amylase Assay Kit provides a solution-based assay featuring the speed, high sensitivity, and convenience required for measuring amylase activity or for screening amylase inhibitors in a high-throughput format. This EnzChek kit contains a starch derivative—the DQ™ starch substrate—that is labeled with BODIPY FL dye to such a degree that the fluorescence is quenched. This substrate is efficiently degraded by amylase; digestion relieves the quenching and yields highly fluorescent fragments. The accompanying increase in fluorescence is proportional to amylase activity and can be monitored with a fluorescence microplate reader or fluorometer, using standard fluorescein filters.

  • Label (Ex/Em): BODIPY FL conjugate (approx. 502/512nm)
  • Kit contains lyophilized substrate, 10X reaction buffer, substrate solvent, a fluorescent standard, and a detailed protocol
  • Sufficient reagents are supplied for 500 assays (using a 100μL assay volume in a 96-well microplate assay format)

Find Fluorescent Substrates for Other Glycosidases

In addition to the EnzChek Ultra Amylase Assay Kit, we offer kits and substrates to measure xylanase, lysozyme, β-galactosidase, and more.

Cell Analysis, Cellular Imaging, Enzyme and Protein Activity Assays, Glycosidase, Neuraminidase, and Related Enzyme Activity, High-Content Screening (HCS), Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature and protect from light.
Detection Method Fluorescence Intensity
For Use With (Application) Ultra Amylase Assay
For Use With (Equipment) Fluorescence Microplate Reader
Product Type EnzChek Assay Kit
Product Line EnzChek
Quantity 1 Kit
Shipping Condition Room Temperature
Substrate Properties Molecular Substrate
Substrate Type Amylase Substrate
Target Enzyme Amylase
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.