Learn More
Invitrogen™ ELF™ 97 Phosphatase Substrate (ELF™ 97 Phosphate), 0.2 μm Filtered
For use with microarray, flow cytometer and fluorescence microscope
Brand: Invitrogen™ E6588
Code : 50
Additional Details : Weight : 0.00525kg
Description
The ELF 97 phosphatase substrate upon hydrolysis produces a bright and photostable yellow-green fluorescent precipitate at the site of enzymatic activity. This fluorescent precipitate has several unique spectral characteristics, including an extremely large Stokes shift that makes it easily distinguishable from endogenous fluorescence.
Cell Analysis, Cell Culture, Cell Viability, Proliferation & Function, Cellular Imaging, Differentiation & Development, Drug Discovery & Development, Enzyme & Protein Activity Assays, IHC Staining & Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Staining & Detection, Immunohistochemistry (IHC), Phosphatase Activity, Phosphatase Activity Assays, Phosphatase Biology, Proteins, Expression, Isolation & Analysis, Stem Cell Analysis, Stem Cell Differentiation, Stem Cell Labeling & Detection, Stem Cell Research, Target & Lead Identification & Validation
Order Info
Shipping Condition: Room Temperature
Specifications
Cell-Permeant | |
5 mM | |
Fluorescent | |
ELF™ | |
Phosphatase Substrate | |
Phosphatases | |
Fluorescence Microscope, Microarray, Flow Cytometer | |
Differentiation |
Yellow-Green | |
Store in freezer (-5 to -30°C) and protect from light. | |
Liquid | |
1 mL | |
Chemical Substrate | |
345⁄530 | |
ELF 97 | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.