missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Deoxyribonuclease I, Alexa Fluor™ 594 Conjugate
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
The bright red fluorescent Alexa Fluor 594 DNase I selectively binds to G-actin (globular).
Cell Analysis, Cell Structure, Cytoskeleton
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Quantity | 5 mg |
| Conjugate | Alexa Fluor 594 |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Excitation/Emission | 590⁄617 |
| Label or Dye | Alexa Fluor™ 594 |
| Label Type | Alexa Fluor Dyes |
| Shipping Condition | Room Temperature |
| Sub Cellular Localization | Actin, Cytoskeleton |
| Form | Lyophilized |
| Product Line | Alexa Fluor |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction