missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CMAC, t-BOC-Leu-Met (7-Amino-4-Chloromethylcoumarin, t-BOC-L-Leucyl-L-Methionine amide)

Product Code. 11594117
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11594117 5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11594117 Supplier Invitrogen™ Supplier No. A6520
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Cleavage produces blue-fluorescence

The CMAC peptidase substrate, t-BOC-Leu-Met can be used to measure peptidase activity in solution or in live cells. The substrate has an excitation/emission maxima approx. 330/403. After cleavage by peptidases, the product produces blue-fluorescence with excitation/emission maxima approx.351/430nm.

Apoptosis, Caspase Activation, Cell Analysis, Cell Viability, Proliferation and Function, Enzyme and Protein Activity Assays, Protease/Peptidase Activity, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Cell-Permeant
Content And Storage Store in freezer (-5 to -30°C).
Detection Method Fluorescence
Form Solid
Quantity 5 mg
Substrate CMAC peptidase substrate
Substrate Properties Peptide-Based Substrate
Substrate Type Peptidase Substrate
Target Enzyme Calpain
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.