missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CandyCane™ Glycoprotein Molecular Weight Standards

Product Code. 10788434
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10788434 400 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10788434 Supplier Invitrogen™ Supplier No. C21852

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

CandyCane glycoprotein molecular weight standards separate into alternating bands of glycosylated and nonglycosylated proteins during polyacrylamide gel electrophoresis

  • System: SDS-PAGE
  • Size Range: 14 to 180 kDa
  • Stain Type: Unstained
  • Ready to Load: No
  • Gel Compatibility: NuPAGE™ Tris-Acetate Gels, NuPAGE™ Bis-Tris Gels, Novex™Tris-Glycine Gels, SDS-PAGE Gels, Novex™Tricine Gels

1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Ready to Load No
Size Range 14 to 180 kDa
Gel Compatibility Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
Quantity 400 μL
Shipping Condition Room Temperature
Product Line CandyCane
Product Type Glycoprotein Molecular Weight Standard
Stain Type Unstained
System Type SDS-PAGE

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.