missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phalloidin Labeling Probes

Product Code. 10125002
Change view
Click to view available options
Color:
Blue
Far-red
Green
Near-infrared
None
Orange
Orange-red
Red
Red-orange
Violet
Yellow
Excitation Wavelength Range:
346/442 nm
405/450 nm
495/518 nm
496/516 nm
496/520 nm
531/554 nm
540/565 nm
555/565 nm
556/570 nm
558/569 nm
578/600 nm
581/609 nm
591/608 nm
632/647 nm
633/647 nm
650/668 nm
663/690 nm
679/702 nm
758/784 nm
None
Dye Type:
Alexa Fluor Plus 555
Alexa Fluor Plus 647
Alexa Fluor™ 350
Alexa Fluor™ 488
Alexa Fluor™ 532
Alexa Fluor™ 546
Alexa Fluor™ 555
Alexa Fluor™ 568
Alexa Fluor™ 594
Alexa Fluor™ 633
Alexa Fluor™ 635
Alexa Fluor™ 647
Alexa Fluor™ 660
Alexa Fluor™ 680
Alexa Fluor™ Plus 405
Alexa Fluor™ Plus 750
BODIPY™
Biotin-XX
FITC (Fluorescein)
Oregon Green™ 488
Phalloidin (unlabeled)
TRITC
Texas Red™
Unit Size:
1mg
300 units
50µg
5g
Each
Pack of 300
23 product options available for selection
Product selection table with 23 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Excitation Wavelength Range Dye Type unitSize
10125002 None None Biotin-XX Each
10174712 Blue 346/442 nm Alexa Fluor™ 350 Each
15932941 Violet 405/450 nm Alexa Fluor™ Plus 405 Each
10125092 Green 495/518 nm Alexa Fluor™ 488 300 units
10730355 Green 496/520 nm Oregon Green™ 488 Each
10615963 Green 496/516 nm FITC (Fluorescein) Each
10184712 Yellow 531/554 nm Alexa Fluor™ 532 Each
10063052 Red-orange 540/565 nm TRITC 300 units
10634053 Orange 556/570 nm Alexa Fluor™ 546 Each
10389132 Orange 555/565 nm Alexa Fluor™ 555 Each
16628105 Orange 555/565 nm Alexa Fluor Plus 555 50µg
10643853 Red 558/569 nm BODIPY™ Each
10135092 Orange-red 578/600 nm Alexa Fluor™ 568 Each
10751144 Red 581/609 nm Alexa Fluor™ 594 Each
10164852 Red 591/608 nm Texas Red™ Each
10053252 Far-red 632/647 nm Alexa Fluor™ 633 5g
10124892 Far-red 633/647 nm Alexa Fluor™ 635 Each
10656353 Far-red 650/668 nm Alexa Fluor™ 647 5g
16400815 Far-red 650/668 nm Alexa Fluor Plus 647 Pack of 300
10402322 Near-infrared 663/690 nm Alexa Fluor™ 660 Each
10656163 Near-infrared 679/702 nm Alexa Fluor™ 680 Each
16316512 Near-infrared 758/784 nm Alexa Fluor™ Plus 750 Each
10718853 None None Phalloidin (unlabeled) 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
23 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10125002 Supplier Invitrogen™ Supplier No. B7474

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Achieve precise and reliable F-actin staining with fluorescent and biotinylated phalloidins. Phalloidin conjugates are widely used in imaging applications to selectively label F-actin in a variety of sample types including fixed and permeabilized cells, tissue sections, and cell-free experiments.

Fluorescent and biotinylated phalloidins are water soluble and bind to filamentous actin (F-actin) with nanomolar affinity, making them convenient probes for labeling, identifying, and quantifying F-actin in cryopreserved tissue sections, fixed and permeabilized cells, and cell-free experiments. Phalloidin conjugates bind similarly to actin from various species, including plants and animals, enabling staining of the cytoskeleton in a wide range of samples.

A variety of phalloidin conjugates for filamentous (F-actin) staining are available, including fluorescent Alexa Fluor and Alexa Fluor Plus phalloidins, along with phalloidins conjugated to classic fluorescent dyes such as BODIPY, fluorescein, and rhodamine. Phalloidin staining is spectrally compatible with other fluorescent stains used in cellular analyses such as GFP/RFP, Qdot nanocrystals, and other Alexa Fluor conjugates and antibodies. Biotin‐XX Phalloidin can be used to visualize actin filaments via fluorescent streptavidin tags or standard enzyme-mediated avidin/streptavidin techniques such as in electron microscopy. Unlabeled phalloidin is available for use as a control in blocking F‐actin staining or in promoting polymerization.

Phalloidin conjugates bind to both large and small actin filaments with similar affinity in a 1:1 stoichiometry between phallotoxin and actin subunits. They do not bind G-actin monomers.

Alexa Fluor and Alexa Fluor Plus phalloidin conjugates for F-actin staining

Fluorescent Alexa Fluor dye conjugates of phalloidin are popular F-actin stains, offering color choices across the full spectral range. These phalloidin conjugates provide researchers with fluorescent probes that are superior in brightness and photostability compared to other spectrally similar conjugates.

Alexa Fluor Plus Phalloidin conjugates retain the same specificity for actin but offer 3-5 times greater sensitivity and brightness compared to the corresponding Alexa Fluor Phalloidin conjugate. This increased brightness is beneficial for challenging F-actin imaging, such as the super‐resolution microscopy methods SIM and STORM, and for reliable staining of actin stress fibers.

Features of phalloidin probes

  • High specificity—binds selectively to F-actin, which allows for precise labeling of actin filaments in fixed cells and cryopreserved tissues
  • Strong affinity—nanomolar binding affinity for F-actin, which ensures stable and reliable actin staining
  • Extensive fluorescent conjugate options—over twenty conjugated varieties of phalloidin
  • Compatibility with fixed samples—typically used with fixed cells and tissues, making them suitable for actin staining in detailed structural studies, immunofluorescence staining, and IHC applications
  • Multiplexing capability—the wide availability of phalloidin conjugates enables their use in combination with other fluorescent probes and antibodies for multiplex imaging. Biotinylated phalloidin can be made use of in downstream streptavidin steps.
  • Quantitative analysis—can be used for quantitative analysis of F-actin distribution and density within cells, aiding in the study of cytoskeletal dynamics. The unlabeled phalloidin can be titrated as a control.
  • Ease of use—staining is straightforward and quick
  • Excellent stability—exhibit good photostability, which is essential for prolonged imaging sessions and time-lapse studies
  • Wide applicability—used for a range of applications, including studying cell morphology, motility, and the effects of drugs on the actin cytoskeleton

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color None
Content And Storage Store in freezer -5°C to -30°C.
Excitation Wavelength Range None
Dye Type Biotin-XX
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer, Confocal Microscope
Quantity 50 Units
Shipping Condition Room Temperature
Label Type Biotin and Other Haptens
Product Type Phalloidin
SubCellular Localization Actin, Cytoskeleton

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.