missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Amplex™ Red/UltraRed Stop Reagent

Product Code. 10093442
Change view
Click to view available options
Quantity:
1 set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10093442 1 set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10093442 Supplier Invitrogen™ Supplier No. A33855

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Designed for use with both the Amplex Red and Amplex UltraRed fluorogenic substrates and assay kits

Amplex Red/UltraRed Stop Reagent provides convenience and control by allowing the fluorescence signal-generating reaction to be terminated at a user-determined time point. enic substrates and assay kits. It is designed to terminate reactions containing up to 0.1 units/mL of horseradish peroxidase (HRP) and 5μM hyrogen peroxide. After addition of the stop reagent, the fluorescence signal remains stable for at least 3 hours.

  • Sensitive biomolecular assays based on hydrogen peroxide–generating enzyme systems linked to peroxidase–mediated oxidation of the fluorogenic Amplex UltraRed or Amplex Red substrates
  • Detection reactions are performed in microplate wells and are initiated by adding fluorogenic Amplex UltraRed or Amplex Red substrate, resulting in a continuous fluorescence increase that proceeds for 30 minutes or more
  • Unknown analyte concentrations are determined by comparing fluorescence intensities measured at a certain time point during the reaction to parallel measurements at same time point on standard samples of known concentration
  • It is critical to ensure that timing of standard and unknown sample measurements is same

Build Your Own Immunoassay, Cell Analysis, Enzyme and Protein Activity Assays, Immunoassays, Lipase and Phopholipase Activity, Oxidase and Peroxidase Activity, Proteins, Expression, Isolation and Analysis, Ready-To-Use Immunoassay

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator at 2°C to 8°C
For Use With (Application) Protein Assays
Product Type Stop Solutions
Form Lyophilized
Product Line Amplex
Quantity 1 set
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.