Learn More
Invitrogen™ Alexa Fluor™ 555-aha-dCTP
Description
The Alexa Fluor™ 555 dyes, used to label nucleotides, are compatible with commonly used microarray scanners, and provide greater signal correlation (R2) values than the spectrally similar Cy™3 and Cy5 dye pair, improving the resolution of two-color microarray gene expression assays to 1.3-fold changes in expression. The exceptionally bright and photostable Alexa Fluor dyes are also essentially insensitive to pH and are highly water soluble.
Array CGH, DNA and RNA Purification and Analysis, DNA Labeling, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Nucleic Acid Labeling and Oligo Synthesis
Order Info
Shipping Condition: Wet Ice
Specifications
Specifications
| Product Type | Labeled Nucleotide |
| Concentration | 1 mM |
| Assay | Microarray Gene Expression Assay |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Label or Dye | Alexa Fluor Dye |
| Labeling Method | Direct Labeling |
| Modification | AHA (5-aminohexylacrylamido) |
| Shipping Condition | Wet Ice |
| Product Line | Alexa Fluor™ |
| Quantity | 50 μL |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.