Learn More
Invitrogen™ Molecular Probes™ aha-dCTP (5-Aminohexylacrylamido-dCTP)
Product Code. 10053432
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
Each
Product Code. Quantity unitSize
10053432 500 μL Each
1 options
Product Code. 10053432 Supplier Invitrogen™ Supplier No. A32768
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Supports two-step technique for improved DNA labeling results
Aha-dCTP (5-aminohexylacrylamido-dCTP) is modified at the C-5 position of cytosine with a unique hexylacrylamide moiety. aha-dCTP can be used to produce amine-modified DNA by conventional enzymatic incorporation methods such as reverse transcription. The amine-modified DNA can then be labeled with any amine-reactive dye to generate labeled nucleic acid hybridization probes. This two-step labeling technique consistently results in a uniform and high degree of DNA labeling that is difficult to obtain by other methods. In addition to this 2mM solution in TE (500μL unit size), it is available as a 50mM solution in TE (50μL unit size, Cat. No. A32769).
DNA and RNA Purification and Analysis, DNA Labeling, Nucleic Acid Labeling and Oligo Synthesis
TRUSTED_SUSTAINABILITY
Specifications
Product Type Nucleotide, aha-dCTP (5-Aminohexylacrylamido-dCTP)
For Use With (Application) DNA and RNA Purification and Analysis, DNA Labeling, Nucleic Acid Labeling and Oligo Synthesis
Quantity 500 μL