missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Molecular Probes™ 4-Bromo A-23187, Free Acid

Product Code. 10124192
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10124192 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10124192 Supplier Invitrogen™ Supplier No. B1494

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For in situ calibrations of fluorescent Ca2+ indicators

  • Suitable to equilibrate intracellular and extracellular Ca2+ concentrations and to permit Mn2+ to enter cell to quench intracellular dye fluorescence
  • Unlike A-23187, brominated A-23187 is essentially nonfluorescent and can be used with UV-excitable calcium indicators like fura-2, indo-1 and quin-2 as well as visible light-excitable indicators, including fluo-4, fluo-3, Calcium Green and rhod-2
  • 4-bromo A-23187 can also be used to equilibrate extracelular and intrcellular Mg2+ concentrations, making it useful for calibrating Mg2+ indicators

Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Quantity 1 mg

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.