Learn More
Neogen Molecular Detection Assay 2
Click to view available options
Quantity:
48 Pack
96 Pack
Unit Size:
1 set
Description
- One assay protocol and fewer steps than most tests reduces repetitive tasks and makes it easy to train employees.
- Ready-to-use and pre-dispensed reagents provide convenience and minimize the chance of human error.
- Color-coded assay tubes by organism and matching software screens allow for better tracking and easy identification.
- The ability to run all assays simultaneously and the flexibility to test up to 96 samples in each run helps you operate more efficiently.
- Real-time amplification and detection process identifies positive results in as early as 15 minutes and allows you to make critical decisions faster.
Specifications
Specifications
| Description | STEC Gene Screen |
| Product Type | Microbiology Media |
| Test Type | Food and Beverage, Environmental Test |
| Quantity | 96 Pack |