missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Molecular Biology Reagents
BIOPHARMA

Product Code. 15343711
Click to view available options
Chemical Name or Material:
Saline Sodium Citrate Buffer
TAE Buffer
Tris Borate EDTA Buffer
Tris Hydrochloride Buffer
pH:
7 ±0.1
7.5 ±0.1
8 ±0.1
8.3 ±0.1
8.4 ±0.1
Unit Size:
Pack of 6
This item is not returnable. View return policy

Product Code. 15343711

Brand: Corning™ 46010CM

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in molecular biology, nucleic acid purification applications

  • Certificate of Analysis available
  • Product can be diluted to obtain the desired concentration

Order Info

Shipped at ambient temperature

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material TAE Buffer
Concentration 10X
pH 8.3 ±0.1
Quantity 6 x 1 L
Sterility Sterile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.