Learn More
Erlab™ Molecode S
Click to view available options
For Use With (Equipment):
Captair Smart Fume Hoods
Unit Size:
Each
Description
- Equipment required by class I of the AFNOR standard NF X 15-211: 2009
- A cell located in the sampling chamber allows automatic detection the saturation of the filter by the solvents
- A second cell is placed in contact with the environment of the laboratory indicates a possible pollution of the ambient air
Specifications
Specifications
| For Use With (Equipment) | Captair Smart Fume Hoods |
| Product Type | Saturation Detection Alarm |
| For Use With (Application) | Detecting Solvents |