Learn More
Moldex™ 8000 Series Half-Mask Respirators Cartridges
Description
- Use with 8910 N95, 8970 R95 or 8940 P100 filters against particulates
- The P100 cartridge can be used for both oil and non-oil based particulates
- 100% PVC-Free™
Specifications
Specifications
| Cartridge Type | Organic Vapor Cartridge |
| Certifications/Compliance | NIOSH |
| Product Type | Organic Vapor Cartridge |
| For Use With (Application) | For protection against organic vapors |
| For Use With (Equipment) | Series 8000 half mask, n95, r95, p100 filter |
| Quantity | 10/Pk. |