Learn More
Moldex™ 8000 Series Half-Mask Facepieces
Product Code. 12332309
Click to view available options
Format:
Small
Medium
Unit Size:
Each
Product Code. 12332309
Brand: Moldex™ 800101
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Soft, durable facepiece confroms to the contours of the face without pressure points to provide long lasting comfort
  • Extra wide surface seals easily and stays sealed even when talking
  • Low profile design provides a large field of vision
  • Facepiece has no gaskets or yoke; fewer parts to maintain and inventory
  • Each cartridge includes a new inhalation valve and features snap-in mount that eliminates mis-threading
Specifications
Certifications/Compliance NIOSH
Connection Type Snap-in gasket
Material Thermoplastic Elastomer
Product Type Half Mask Facepiece
Format Small