Learn More
Honeywell™ Molded Cup Facemask 5211 XL-FFP2 NR D with Valve and Full-Face Seal
Product Code. 12601455
Change view
Click to view available options
:
Size: X-Large
Product Code.
12601455 Size: X-Large
1 options
Product Code. 12601455 Supplier Honeywell™ Supplier No. 1005120
Only null left
Add to basket
Edge
Add to basket
Rigid shell facemask with an ultra-soft flange seal for comfort and fit and a high-performance exhalation valve for improved protection.
  • Rigid shell: shape maintained even in moistened and heavy loading environments.
  • Ultra-soft hypoallergenic Willtech™ flange seal with absorption properties and a tight-sealing film for an exceptional comfort and a perfect fit.
  • Preformed and colored nosebridge to save time to perfectly adjust the mask and to instantly identify its protection level.
  • Large internal volume to easily communicate and breathe.
  • Shielded, high performance exhalation valve with an exclusive design (butterfly opening-closing system) for improved protection, comfort, and decreased fatigue.
  • Adjustable braids to easily and efficiently fit mask.
  • Lightweight respirator (26 g) to reduce fatigue.
  • No latex, no silicone, no PVC.
  • Dolomite tested.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EC Category PPE: 3, QA: ISO 9001/2000, EC Attestation, EN 149:2001+A1:2009
Disposable Single-use
Product Type Rigid Shell
Size X-Large
With Exhalation Valve Yes
For Use With (Application) Agriculture, automotive, construction, food, iron/steel, mining, textile, wood industries
Includes Valve, adjustable braids, full-face seal
Latex Free Yes
Quantity 5/Box, 50/Pk.