missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MOK VHH-8His-Cys-tag Recombinant Alpaca Monoclonal Antibody (SAA1290)

Product Code. 30131089
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30131089 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30131089 Supplier Invitrogen™ Supplier No. MA554670

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Alpaca Recombinant Monoclonal Antibody

Compatible with Direct ELISA.

MOK is able to phosphorylate several exogenous substrates and to undergo autophosphorylation. It negatively regulates cilium length in a cAMP and mTORC1 signaling-dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOK VHH-8His-Cys-tag
Applications ELISA, Immunoprecipitation
Classification Recombinant Monoclonal
Clone SAA1290
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene MOK
Gene Accession No. Q9UQ07
Gene Alias MAPK/MAK/MRK overlapping kinase; MAPK/MAK/MRK/ overlapping kinase; MOK; MOK protein kinase; Rage; RAGE1; RAGE-1; renal cell carcinoma antigen; renal tumor antigen; renal tumor antigen 1; Serine/threonine kinase 30; soluble RAGE; STK30; T/STK 30
Gene Symbols MOK
Host Species Alpaca
Immunogen Recombinant Human RAGE-1/MOK.
Purification Method Metal-chelate Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5891
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype VHH
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.