missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOK Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23926-1-AP-20UL
This item is not returnable.
View return policy
Description
Able to phosphorylate several exogenous substrates and to undergo autophosphorylation.Specifications
| MOK | |
| Polyclonal | |
| Unconjugated | |
| MOK | |
| MOK, MOK protein kinase, RAGE, RAGE 1, RAGE1, renal tumor antigen, Renal tumor antigen 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5891 | |
| -20°C | |
| Liquid |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.46 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UQ07 | |
| MOK | |
| MOK Fusion Protein Ag20849 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction