missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MOK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102485
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MOK.
MOK is able to phosphorylate several exogenous substrates and to undergo autophosphorylation. It negatively regulates cilium length in a cAMP and mTORC1 signaling-dependent manner.
Specifications
| MOK | |
| Polyclonal | |
| Unconjugated | |
| MOK | |
| MAPK/MAK/MRK overlapping kinase; MAPK/MAK/MRK/ overlapping kinase; MOK; MOK protein kinase; Rage; RAGE1; RAGE-1; renal cell carcinoma antigen; renal tumor antigen; renal tumor antigen 1; Serine/threonine kinase 30; soluble RAGE; STK30; T/STK 30 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 26448, 362787, 5891 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UQ07, Q9WVS4 | |
| MOK | |
| A synthesized peptide derived from human MOK(Accession Q9UQ07), corresponding to amino acid residues L135-L185. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction