missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MOK Polyclonal Antibody
GREENER_CHOICE

Product Code. 16528771
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16528771 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16528771 Supplier Invitrogen™ Supplier No. PA595928

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MKNYKAIGKI GEGTFSEVMK MQSLRDGNYY ACKQMKQRFE SIEQVNNLRE IQALRRLNPH PNILMLHEVV FDRKSGSLAL ICELMDMNIY ELIRGRRYPL SEKKIMHYMY QLCKSLDHIH RNGIFHRDVK PENILIKQDV LKLGDFGSCR SVYSKQPYTE YISTRWYRAP ECLLTDGFYT YKMDLWSAGC VFYEIASLQP LFPGVNELDQ ISKIHDVIGT PAQKILTKFK; Positive Samples: Mouse liver; Cellular Location: Cytoplasm.

MOK is able to phosphorylate several exogenous substrates and to undergo autophosphorylation. It negatively regulates cilium length in a cAMP and mTORC1 signaling-dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOK
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MOK
Gene Accession No. Q9UQ07, Q9WVS4
Gene Alias MAPK/MAK/MRK overlapping kinase; MAPK/MAK/MRK/ overlapping kinase; MOK; MOK protein kinase; Rage; RAGE1; RAGE-1; renal cell carcinoma antigen; renal tumor antigen; renal tumor antigen 1; Serine/threonine kinase 30; soluble RAGE; STK30; T/STK 30
Gene Symbols MOK
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human MOK (NP_0550411).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26448, 362787, 5891
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.