missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mohawk homeobox Antibody (OTI6B12), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72758FR
This item is not returnable.
View return policy
Description
Mohawk homeobox Monoclonal specifically detects Mohawk homeobox in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Mohawk homeobox | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| MKX | |
| Full length recombinant protein of human Mohawk homeobox (NP_775847) produced in E.coli. | |
| 0.1 mL | |
| 283078.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI6B12 | |
| Western Blot, Immunohistochemistry | |
| C10orf48, homeobox protein Mohawk, IFRX, Iroquois family related homeodomain protein, iroquois homeobox protein-like 1, IRXL1chromosome 10 open reading frame 48, MGC39616, mohawk homeobox | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction