Learn More
Invitrogen™ MOGAT3 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5103518
Product Code. 16699964
  • £440.00 / 100µL
Description
Antibody detects endogenous levels of total MOGAT3.
Catalyzes the formation of diacylglycerol from 2- monoacylglycerol and fatty acyl-CoA. Also able to catalyze the terminal step in triacylglycerol synthesis by using diacylglycerol and fatty acyl-CoA as substrates. Has a preference toward palmitoyl-CoA and oleoyl-CoA. May be involved in absorption of dietary fat in the small intestine by catalyzing the resynthesis of triacylglycerol in enterocytes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MOGAT3
Polyclonal
Unconjugated
MOGAT3
2-acylglycerol O-acyltransferase 3; acyl coenzyme A:monoacylglycerol acyltransferase 3; acyl-CoA:monoacylglycerol acyltransferase 3; DC7; DGAT2L2; DGAT2L7; Diacylglycerol acyltransferase 2-like protein 7; diacylglycerol O-acyltransferase candidate 7; hDC7; MGAT3; MOGAT3; Monoacylglycerol O-acyltransferase 3; UNQ9383/PRO34208
Rabbit
Affinity Chromatography
RUO
346606, 685560
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q86VF5
MOGAT3
A synthesized peptide derived from human MOGAT3(Accession Q86VF5), corresponding to amino acid residues H302-I341.
100 μL
Primary
Human, Rat