missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOGAT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21286-1-AP-150UL
This item is not returnable.
View return policy
Description
Dietary fat absorption from the small intestine is facilitated by acyl-CoA:monoacylglycerol transferase (MOGAT; EC 2.3.1.22) and acyl-CoA:diacylglycerol acyltransferase (DGAT; see MIM 604900) activities. MOGAT catalyzes the joining of monoacylglycerol and fatty acyl-CoAs to form diacylglycerol (Yen and Farese, 2003 ).Specifications
| MOGAT2 | |
| Polyclonal | |
| Unconjugated | |
| MOGAT2 | |
| DC5, DGAT2L5, hDC5, hMGAT2, MGAT2, MOGAT2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 233549, 681211, 80168 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3SYC2, Q80W94 | |
| Mogat2 | |
| MOGAT2 Fusion Protein Ag15806 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur