missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOGAT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21573-1-AP-20UL
This item is not returnable.
View return policy
Description
Catalyzes the formation of diacylglycerol from 2-monoacylglycerol and fatty acyl-CoA. Probably not involved in absorption of dietary fat in the small intestine.Specifications
| MOGAT1 | |
| Polyclonal | |
| Unconjugated | |
| MOGAT1 | |
| DC2, DGAT2L, DGAT2L1, hDC2, MGAT1, MOGAT1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 116255, 363261, 68393 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q91ZV4, Q96PD6 | |
| Mogat1 | |
| MOGAT1 Fusion Protein Ag16109 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction