missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mogamulizumab Humanized Recombinant Human Monoclonal Antibody

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA541932
This item is not returnable.
View return policy
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Mogamulizumab is a humanized monoclonal antibody (mAb) directed against CC chemokine receptor 4 (CCR4) for the treatment of Mycosis Fungoides (MF) and Sezary Syndrome (SS), the most common subtypes of cutaneous T-cell lymphoma. Cutaneous T-cell lymphomas occur when certain white blood cells, called T cells, become cancerous; these cancers typically affect the skin, causing various types of skin lesions 8. On August 8 2018, the U. S. Food and pharmaceutical Administration (FDA) approved mogamulizumab injection (also known as Poteligeo) for intravenous use for the treatment of adult patients with relapsed or refractory mycosis fungoides (MF) or Sezary syndrome (SS) after at least one prior systemic therapy 7. Mogamulizumab is derived from Kyowa Hakko Kirin's POTELLIGENT (™) technology, which produces antibodies with enhanced antibody-dependent cell-mediated cytotoxicity (ADCC) activity. Approval in Japan was granted on April 30 2012 by the Japanese Ministry of Health, Labor and Welfare for patients with relapsed or refractory CCR4-positive adult T-cell leukemia-lymphoma 2.
Specifications
| Mogamulizumab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| AMG-761; KW-0761 | |
| Human CCR4/CD194. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
| ELISA, Flow Cytometry | |
| 2.66 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Human | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction