missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mogamulizumab Chimeric Recombinant Mouse Monoclonal Antibody (KW-0761 (Mogamulizumab))

Mouse Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551767
This item is not returnable.
View return policy
Description
This antibody is specific for human CCR4, a chemokine receptor that is preferentially expressed by Th2 and regulatory T (Treg) cells. CCR4 is also detected on basophils, platelets, monocytes and brain microvascular and coronary artery endothelial cells.
Mogamulizumab is a humanized monoclonal antibody (mAb) directed against CC chemokine receptor 4 (CCR4) for the treatment of Mycosis Fungoides (MF) and Sezary Syndrome (SS), the most common subtypes of cutaneous T-cell lymphoma. Cutaneous T-cell lymphomas occur when certain white blood cells, called T cells, become cancerous; these cancers typically affect the skin, causing various types of skin lesions 8. On August 8 2018, the U. S. Food and pharmaceutical Administration (FDA) approved mogamulizumab injection (also known as Poteligeo) for intravenous use for the treatment of adult patients with relapsed or refractory mycosis fungoides (MF) or Sezary syndrome (SS) after at least one prior systemic therapy 7. Mogamulizumab is derived from Kyowa Hakko Kirin's POTELLIGENT (®) technology, which produces antibodies with enhanced antibody-dependent cell-mediated cytotoxicity (ADCC) activity. Approval in Japan was granted on April 30 2012 by the Japanese Ministry of Health, Labor and Welfare for patients with relapsed or refractory CCR4-positive adult T-cell leukemia-lymphoma 2.
Specifications
| Mogamulizumab Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Mouse | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG2a κ |
| Functional Assay | |
| KW-0761 (Mogamulizumab) | |
| Unconjugated | |
| AMG-761; KW-0761 | |
| This antibody was prepared by humanization of a mouse anti-CCR4 mAb. | |
| 200 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction