missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOG Antibody (CL2858), Novus Biologicals™
Mouse Monoclonal Antibody
£265.00 - £458.00
Specifications
| Antigen | MOG |
|---|---|
| Clone | CL2858 |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000, Immunohistochemistry-Frozen |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18605295
|
Novus Biologicals
NBP2-46634-25ul |
25 μL |
£265.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18669955
|
Novus Biologicals
NBP2-46634 |
0.1 mL |
£458.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MOG Monoclonal antibody specifically detects MOG in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)Specifications
| MOG | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000, Immunohistochemistry-Frozen | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Q16653 | |
| 4340 | |
| IgG1 | |
| Protein A purified |
| CL2858 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Cellular Markers | |
| PBS (pH 7.2), 40% Glycerol | |
| MGC26137, MOG Ig-AluB, MOGIG2, myelin oligodendrocyte glycoprotein, myelin-oligodendrocyte glycoprotein | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence ALVGDEVELPCRISPGKNATGMEVGWYRPPFSRVVHLYRNGKDQDGDQAPEYRGRTELLKDAIGEGKVTLRIRNVRFSDEGGFTCFFRDHSYQEEAAMELKVEDPFYWVSPG | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title