missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MOG Monoclonal antibody specifically detects MOG in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | MOG |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 013 |
| Conjugate | Alexa Fluor 532 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | MGC26137, MOG Ig-AluB, MOGIG2, myelin oligodendrocyte glycoprotein, myelin-oligodendrocyte glycoprotein |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human MOG (NP_996532.2; Gly 30-Tyr 149). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?