missing translation for 'onlineSavingsMsg'
Learn More

Mofegiline hydrochloride, MedChemExpress

Product Code. 16499657
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499657 5 mg 5mg
16497118 10 mg 10mg
16453157 50 mg 50mg
16417146 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16499657 Supplier Medchem Express Supplier No. HY16677A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mofegiline hydrochloride (MDL72974A) is a potent and selective enzyme-activated irreversible inhibitor of MAO-B; shows marked selectivity for the B form (IC50 = 680 and 3.6 nM for MAO-A and MAO-B).

Mofegiline hydrochloride (MDL72974A) is a potent and selective enzyme-activated irreversible inhibitor of MAO-B

Specifications

Chemical Name or Material Mofegiline hydrochloride
CAS 120635-25-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C11H14ClF2N
Quantity 5 mg
Synonym MDL72974A
Solubility Information DMSO : 110 mg/mL (470.71 mM; Need ultrasonic) ∣H2O : 25 mg/mL (106.98 mM; Need ultrasonic)
SMILES FC1=CC=C(CC/C(CN)=C(F)/[H])C=C1.[H]Cl
Molecular Weight (g/mol) 233.69
Formula Weight 233.69
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.