missing translation for 'onlineSavingsMsg'
Learn More

Moexipril hydrochloride, MedChemExpress

Product Code. 16408468
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16408468 10 mg 10mg
16498266 100 mg 100mg
16434347 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16408468 Supplier Medchem Express Supplier No. HYB0378A10mg
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Moexipril hydrochloride (RS-10085) is an orally active inhibitor of angiotensin-converting enzyme (ACE), and becomes effective by being hydrolyzed to moexiprila (hydrochloride). Moexipril hydrochloride exhibits antihypertensive and neuroprotective effects-.

Moexipril hydrochloride (RS-10085) is an orally active inhibitor of angiotensin-converting enzyme (ACE), and becomes effective by being hydrolyzed to moexiprila (hydrochloride). Moexipril hydrochloride exhibits antihypertensive and neuroprotective effects-.

Specifications

Chemical Name or Material Moexipril hydrochloride
CAS 82586-52-5
Color White
Purity Grade Notes Research
Molecular Formula C27H35ClN2O7
Quantity 10 mg
Synonym RS-10085
Solubility Information DMSO : 250 mg/mL (467.26 mM; Need ultrasonic) ∣H2O : 100 mg/mL (186.91 mM; Need ultrasonic)
SMILES O=C([C@H]1N(C([C@@H](N[C@H](C(OCC)=O)CCC2=CC=CC=C2)C)=O)CC3=C(C=C(OC)C(OC)=C3)C1)O.Cl
Molecular Weight (g/mol) 535.03
Formula Weight 535.03
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.