missing translation for 'onlineSavingsMsg'
Learn More

Moexipril Diketopiperazine, TRC

Product Code. 30470120
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30470120 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30470120 Supplier Toronto Research Chemicals Supplier No. TRCM48534010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-ethyl 2-{(3S,11aS)-8,9-dimethoxy-3-methyl-1,4-dioxo-3,4-dihydro-1H-pyrazino[1,2-b]isoquinolin-2(6H,11H,11aH)-yl}-4-phenylbutanoate
CAS 103733-51-3
Molecular Formula C27 H32 N2 O6
Quantity 10 mg
Synonym (αS,3S,11aS)-1,3,4,6,11,11a-Hexahydro-8,9-dimethoxy-3-methyl-1,4-dioxo-α-(2-phenylethyl)-2H-pyrazino[1,2-b]isoquinoline-2-acetic Acid Ethyl Ester, [3S-[2(R*),3α,11aβ]]-1,3,4,6,11,11a-Hexahydro-8,9-dimethoxy-3-methyl-1,4-dioxo-α-(2-phenylethyl)-2H-pyrazino[1,2-b]isoquinoline-2-acetic Acid Ethyl Ester, PD 114009, Moexipril USP Related Compound B
InChI Formula InChI=1S/C27H32N2O6/c1-5-35-27(32)21(12-11-18-9-7-6-8-10-18)29-17(2)25(30)28-16-20-15-24(34-4)23(33-3)14-19(20)13-22(28)26(29)31/h6-10,14-15,17,21-22H,5,11-13,16H2,1-4H3
SMILES CCOC(=O)C(CCc1ccccc1)N2C(C)C(=O)N3Cc4cc(OC)c(OC)cc4CC3C2=O
IUPAC Name ethyl 2-(8,9-dimethoxy-3-methyl-1,4-dioxo-3,6,11,11a-tetrahydropyrazino[1,2-b]isoquinolin-2-yl)-4-phenylbutanoate
Molecular Weight (g/mol) 480.55
Formula Weight 480.226
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.