missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Moesin Recombinant Rabbit Monoclonal Antibody (24GB265)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557486
This item is not returnable.
View return policy
Description
Moesin Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Moesin, a member of the talin-4. 1 superfamily, is a linking protein of the submembraneous actin cytoskeleton. It is expressed in macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. It is involved in cell adhesion, migration, and organization of cell surface structures.
Specifications
| Moesin | |
| Recombinant Monoclonal | |
| Unconjugated | |
| MSN | |
| C78546; epididymis luminal protein 70; HEL70; Membrane-organizing extension spike protein; MGC137877 protein; moesin; MSN | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17698, 4478, 81521 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB265 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35763, P26038, P26041 | |
| MSN | |
| A synthesized peptide derived from human Moesin (aa 460-480). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction