missing translation for 'onlineSavingsMsg'
Learn More

Moesin Antibody (MSN/491) - Azide and BSA Free, Novus Biologicals™

Product Code. 18420432 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420432 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420432 Supplier Novus Biologicals Supplier No. NBP2346850.1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Moesin Monoclonal antibody specifically detects Moesin in Human, Rat samples. It is validated for Western Blot, Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Moesin
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone MSN/491
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1 μg/mL, Simple Western, Flow Cytometry 0.5-1 μg/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 0.5-1 μg/mL, Immunohistochemistry-Paraffin 0.5-1 μg/mL, CyTOF-ready
Formulation 10 mM PBS
Gene Alias Membrane-organizing extension spike protein, moesin
Host Species Mouse
Immunogen Recombinant full-length human Moesin protein (Uniprot: P26038)
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4478
Target Species Human, Rat
Content And Storage Store at -20°C to -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.