Learn More
OHAUS™ Module Block
OHAUS™ Module Block features close contact of tubes-to-walls allowing for maximum heat retention and is available with or without a thermometer well.
£123.00 - £277.00
Product Code | Brand | Holds | Includes | No. of Places | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Includes | No. of Places | Price | Quantity & Availability | |||
15722579
|
OHAUS™
30400160 |
0 Tubes x 1 Block | Without Thermometer Well | 0 |
£123.00
Each |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||
15782579
|
OHAUS™
30400166 |
0 Tubes x 2 Block | Without Thermometer Well | 0 |
£177.00
Each |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||
15732579
|
OHAUS™
30400161 |
Cuvettes | With Thermometer Well | 2 |
£145.00
Each |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||
15752609
|
OHAUS™
30400193 |
2 x 50 mL Tubes, 3 x 15 mL Tubes, 4 x 1.5 mL Tubes | With Thermometer Well | 9 |
£123.00
Each |
In Stock
Log in to see stock. |
|||
15782569
|
OHAUS™
30400156 |
3 x 25 mm Tubes, 5 x 12 to 13 mm Tubes, 6 x 6 mm Tubes | With Thermometer Well | 14 |
£123.00
Each |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||
15792579
|
OHAUS™
30400167 |
35 mm Tubes | With Thermometer Well | 4 |
£123.00
Each |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||
15772569
|
OHAUS™
30400155 |
25 mm Tubes | With Thermometer Well | 6 |
£123.00
Each |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||
15762569
|
OHAUS™
30400154 |
20 mm Tubes | With Thermometer Well | 8 |
£123.00
Each |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||
15772609
|
OHAUS™
30400195 |
17 to 18 mm Tubes | With Thermometer Well | 12 |
£123.00
Each |
In Stock
Log in to see stock. |
|||
15772579
|
OHAUS™
30400165 |
12 to 13 mm Tubes | Without Thermometer Well | 16 |
£123.00
Each |
In Stock
Log in to see stock. |
|||
15732609
|
OHAUS™
30400191 |
2 mL Tubes | With Thermometer Well | 20 |
£150.00
Each |
In Stock
Log in to see stock. |
|||
15742609
|
OHAUS™
30400192 |
2 mL Corning Tubes | With Thermometer Well | 20 |
£123.00
Each |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||
15742579
|
OHAUS™
30400162 |
11.5/1.5 mL Micta | With Thermometer Well | 20 |
£123.00
Each |
In Stock
Log in to see stock. |
|||
15742569
|
OHAUS™
30400152 |
12 to 13 mm Tubes | With Thermometer Well | 20 |
£123.00
Each |
In Stock
Log in to see stock. |
|||
15762609
|
OHAUS™
30400194 |
Microtubes | With Thermometer Well | 21 |
£123.00
Each |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||
15792569
|
OHAUS™
30400157 |
0.5 mL Microtaper | With Thermometer Well | 30 |
£123.00
Each |
In Stock
Log in to see stock. |
|||
15752579
|
OHAUS™
30400163 |
250μL Tubes/6 mm Tubes | With Thermometer Well | 30 |
£123.00
Each |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||
15702579
|
OHAUS™
30400158 |
6 mm Tubes | With Thermometer Well | 30 |
£123.00
Each |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||
15722589
|
OHAUS™
30400170 |
0.2 mL Microtubes | Without Thermometer Well | 64 |
£134.00
Each |
In Stock
Log in to see stock. |
|||
15712589
|
OHAUS™
30400169 |
0.2 mL Microtubes | Without Thermometer Well | 80 |
£155.00
Each |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||
15732589
|
OHAUS™
30400171 |
0.2 mL Microtubes | With Thermometer Well | 96 |
£277.00
Each |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||
Descripción
- Block heater accessories include modular blocks to accommodate almost any sample
- Modular blocks feature close contact of tubes-to-walls allowing for maximum heat retention
- Most blocks feature a thermometer well for measuring block temperature
- Low temperature covers help to reduce the air flow for additional temperature stability
Especificaciones
Anodized Aluminum | |
Block |