Learn More
New Pig™ Modular Spill Deck 4 Drum
Product Code. 12941405
Encompass_Preferred
Product Code. 12941405 Supplier New Pig™ Supplier No. PAK531
Light duty polyethylene with UV inhibitors 28 kg 1320mm x 1320mm x 150mm 84L sump capacity sump white grate black