missing translation for 'onlineSavingsMsg'
Learn More

Ohaus™ Block

Material: Anodized Aluminum; For Use With: 96-well or 384-well titer plate

Brand:  Ohaus™ 30400164

Additional Details : Weight : 1.60000kg

Product Code. 15762579

  • £256.00 / Each
Estimated Shipment: 06-08-2024
to see stock.
Add to basket

Alert:

To avoid possible electrical hazard, DO NOT fill well or block with water or other liquids. Units are designed as a dry bath/incubator.


Description

Description

  • Double block
  • Well depth: 13.5mm
  • Fits 2/4/6 block dry block heaters
  • Recessed well for better stability, flat surface for flat and round bottom plates
  • Dimension: 15.2L x 9.5W x 5.7cm H (6 x 3.75 x 2.25 in.)
Specifications

Specifications

Module Block Titer Plate
Anodized Aluminum
Titer plate
6 in.
2.25 in.
0.53 in.
96-well or 384-well titer plate
Block
2 Block
15.2 cm
5.7 cm
1.35 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers