missing translation for 'onlineSavingsMsg'
Learn More

OHAUS™ Modular Blocks and Accessories

Constructed from solid anodized aluminum block.

Brand:  OHAUS™ 30400168

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15702589

  • £142.00 / Each
Estimated Shipment: 04-06-2024
to see stock.
Add to basket

Alert:

To avoid possible electrical hazard, DO NOT fill well or block with water or other liquids. Units are designed as a dry bath/incubator.


Description

Description

Modular blocks are sold separately from heaters.

  • Close contact of tubes-to-block-walls allow for maximum heat retention
  • Each block has a thermometer well for measuring block temperature

Specifications

Specifications

Module Block 50 mL Conical Centrifuge
Anodized Aluminum
Accessory, Block
1 Block
9.5 cm
5.1 cm
4.76 cm
2.9 cm
Dry block heater
5
50mL Tube
3.75 in.
2 in.
1.87 in.
1.14 in.
Recommendations

Recommendations

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers