Learn More
Thermo Scientific™ Modifying Enzymes
Shop All Thermo Scientific ProductsDescription
Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.
High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.
Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.
High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.
Features
- Convenient stand-alone enzymes and buffers for workflow optimization
- No animal-origin components in final product composition
- Reliable and reproducible results with rigorous quality testing
- Scalable and available as OEM products
Applications
- CRISPR/Cas-based genome editing
- Gene expression
- Generation of pluripotent cells
Specifications
Specifications
| Concentration | 50 U/μL |
| For Use With (Application) | mRNA Capping |
| Content And Storage | 50 U/μL Vaccinia mRNA 2'-O-Methyltransferase, 400 μL Store at -20°C. |
| Enzyme | Methyltransferase |
| Compatible Buffer | 10X Capping Buffer |
| Quantity | 20,000 U |
| Product Type | Vaccinia mRNA Cap 2'-O-Methyltransferase |
| Research Category | RNA Research |
| Form | Liquid |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.