missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Modified Harris Hematoxylin
Shop All Epredia Products
Click to view available options
Quantity:
1 pt.
1 gal.
Unit Size:
Each
Pack of 4
Description
- Formulation contains no mercury
- More stable than conventional Harris Hematoxylin
Specifications
Specifications
| Quantity | 1 gal. |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Carcinogenicity Category 1A
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H350-May cause cancer.
- P201-Obtain special instructions before use.
- P362-Take off contaminated clothing.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash face, hands, and anyexposed skin thoroughly after handling.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : hematoxylin, sodiumiodate, ethyl alcohol, methyl alcohol, diazolidinyl urea, acetic acid, aluminium
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction